Acacetin
Glass bottle, 1 g
Catalog # ACRO465230010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- Bioactive
Synonyms: 5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-one, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxy-5,7-dihydroxyflavone, 5,7-Dihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyrone, Linarigenin, Buddleoflavonol, 4'-Methoxyapigenin
- CAS Number:480-44-4
- Formula:C₁₆H₁₂O₅
- MDL Number:MFCD00016936
- Molecular Weight:284.27 g/mol
- Melting Pt:260…265 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to light yellow to light brown Powder or crystals
- Infrared spectrum:Conforms
- Water:≤3 %
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



