Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OBJECTIVE IOS LWD U-PLAN F PH 20X/0.45 1 * 1 items
Catalog # OPTKM-1177
Supplier:
OPTIKA
Expand image
OBJECTIVE IOS LWD U-PLAN F PH 20X/0.45 1 * 1 items
Catalog # OPTKM-1177
Supplier:
OPTIKA