The Whitley Internal HEPA Filtration System achieves levels of atmospheric cleanliness inside our workstations exceeding the requirements of ISO 14644 (Class 3). Key features: - ,,Removable Front with Oval Ports and Sleeves. Permits thorough cleaning 1 * 1 Items
Catalog # DOWIA07070
Supplier: DON WHITLEY SCIENTIFIC
Frequently Bought Together