Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
portable emergency eye, face and partial body shower, capacity 15 l, unheated 1 * 1 items
Catalog # BROEH7170011000303
Supplier:
BROEN-LAB
Expand image
portable emergency eye, face and partial body shower, capacity 15 l, unheated 1 * 1 items
Catalog # BROEH7170011000303
Supplier:
BROEN-LAB