Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Kirjaudu sisään
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FLASH POINT REF. MATS 1 * 200 mL
Catalog # CANN9727-A15
Supplier:
CANNON INSTRUMENT
Expand image
FLASH POINT REF. MATS 1 * 200 mL
Catalog # CANN9727-A15
Supplier:
CANNON INSTRUMENT