Silica PLOT Capillary GC Column, L x I.D. 30 m x 0.53 mm, average thickness 6 mum 1 * 1 items
29758-U
: Merck
Silica PLOT Capillary GC Column, L x I.D. 30 m x 0.53 mm, average thickness 6 mum 1 * 1 items
29758-U
: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection