Order Entry
Finland
ContactUsLinkComponent
SLB(R)-1ms Capillary GC Column, L x I.D. 10 m x 0.10 mm, df 0.10 mum 1 * 1 items
Catalog # 29724-U
Supplier:  Merck
SLB(R)-1ms Capillary GC Column, L x I.D. 10 m x 0.10 mm, df 0.10 mum 1 * 1 items
Catalog # 29724-U
Supplier:  Merck
Frequently Bought Together