Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hotplate with Ceran 500 heating surface 580x430 mm, 5700W, 3x400V+PE, 50...500C 1 * 1 items
Catalog # GEST44A-ON
Supplier:
GESTIGKEIT HARRY
Expand image
Hotplate with Ceran 500 heating surface 580x430 mm, 5700W, 3x400V+PE, 50...500C 1 * 1 items
Catalog # GEST44A-ON
Supplier:
GESTIGKEIT HARRY