Order Entry
Finland
ContactUsLinkComponent
Materials and dialysis tubing are provided to perform procedures involving diffusion, dialysis, water potential and to determine osm otic potential. 1 * 1 KIT
Catalog # EDVO281
Supplier:  EDVOTEK
Materials and dialysis tubing are provided to perform procedures involving diffusion, dialysis, water potential and to determine osm otic potential. 1 * 1 KIT
Catalog # EDVO281
Supplier:  EDVOTEK
Frequently Bought Together