Order Entry
Finland
ContactUsLinkComponent
Transformation is of central importance in molecular cloning since it allows for the selection, propagation, expression and purifica tion of a gene. Positive selection for cells containing plasmid DNA is accomplished by antibiotic growth selection. 1 * 1 KIT
Catalog # EDVO224
Supplier:  EDVOTEK
Transformation is of central importance in molecular cloning since it allows for the selection, propagation, expression and purifica tion of a gene. Positive selection for cells containing plasmid DNA is accomplished by antibiotic growth selection. 1 * 1 KIT
Catalog # EDVO224
Supplier:  EDVOTEK
Frequently Bought Together