CapLock™ clips
CapLock™ clip, Yellow
Catalog # SIMPT330-75CLY
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
- Ideal for shipping samples
- Suitable for 5 ml tubes with snap cap
Specifications
- Colour:Yellow
- Description:CapLock™ clip
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



