Order Entry
Finland
Orders LinkContactUsLinkComponent
CapLock™ clips
CapLock™ clips
CapLock™ clip, Green
  SIMPT330-75CLG
CapLock™ clips
  SIMPT330-75CLG
CapLock™ clips
CapLock™ clips
CapLock™ clip, Green
42,80 €
1 * 100 Items
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
  • Ideal for shipping samples
  • Suitable for 5 ml tubes with snap cap
  • Colour:
    Green
  • Description:
    CapLock™ clip
  • Seq:
    66