Order Entry
Finland
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA

Affinity chromatography columns, HiTrap™ MabSelect™ PrismA

HiTrap™ MabSelect™ PrismA, PP, 60 µm, 16×25 mm, 5 ml, <20 ml/min
Catalog # 17-5498-53
Supplier:   Cytiva
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA
Catalog # 17-5498-53
Supplier:  Cytiva
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA
Affinity chromatography columns, HiTrap™ MabSelect™ PrismA

HiTrap™ MabSelect™ PrismA, PP, 60 µm, 16×25 mm, 5 ml, <20 ml/min

Price
952,00 €
Log in for availability
1 * 1 Items
Product Details & Documents

These columns are prepacked with MabSelect™ PrismA protein A chromatographyn resin that has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs. These columns are suitable for purifying antibodies with affinity chromatography in research and process development.

  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH
  • Convenient HiTrap™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system

MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.

The columns are made of biocompatible PP that does not interact with biomolecules.

Caution: Note that HiTrap™ columns cannot be opened or repacked.

Specifications
  • Flow rate:
    <20 ml/min
  • Particle Size:
    60 µm
  • Capacity:
    5 ml
  • Description:
    HiTrap™ MabSelect™ PrismA
  • Seq:
    03
  • Column hardware:
    PP
  • Bed dimensions:
    16×25 mm
  • Cat. No.:
    17-5498-53
  • Max. pressure:
    5 bar
  • Binding capacity:
    ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • Storage:
    20% ethanol, 2 to 8 °C
  • Temperature limit:
    40 °C
  • pH range:
    3 – 12