Order Entry
Finland
ContactUsLinkComponent
TIC-Ex pipette tips for the expulsion of TIC during the sample preparation of TOC tests, 100 pieces within a PE-bag, in a box 1 * 100 items
Catalog # MANA916998
Supplier:  MACHEREY-NAGEL
undefined
TIC-Ex pipette tips for the expulsion of TIC during the sample preparation of TOC tests, 100 pieces within a PE-bag, in a box 1 * 100 items
Catalog # MANA916998
Supplier:  MACHEREY-NAGEL
Frequently Bought Together