Order Entry
Finland
Orders LinkContactUsLinkComponent
Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
P12×100M, 10 - 100 µl, ±0,25 - ±0,80, ≤0,14 - ≤0,25 µl, Light pink, D200 / DF100
Catalog # 613-6052
Supplier:   GILSON
Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Catalog # 613-6052
Supplier:  GILSON
Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
P12×100M, 10 - 100 µl, ±0,25 - ±0,80, ≤0,14 - ≤0,25 µl, Light pink, D200 / DF100
Price
1 299,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
PIPETMAN® M Connected is a Bluetooth®-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes. Requiring virtually no pipetting forces to aspirate and dispense samples, PIPETMAN M Connected helps reduce pipetting fatigue and increase pipetting efficiency. The pipette interacts in real-time with TRACKMAN® Connected making planning and executing pipetting protocols faster, easier, and more reliable.
  • Simple and intuitive motorized pipette
  • Ultimate consistency
  • Connect to Gilson Apps
With PipetteScope®, you’ll get access to service calibration, pipette activity, and useful resources for your pipette in one location. With PIPETMAN M Connected Software, create custom protocols from 11 different pipetting tasks and sync up to ten custom protocols into your pipette. TRACKMAN Connected check PIPETMAN M Connected calibration status and guides you through the execution of your protocol by displaying where, when, and how much to pipette with real-time interactions and automatic volume adjustments. At the end of the protocol, the application automatically generates a report detailing each of the steps performed with related pictures (e.g. for cells counts...) and notes taken during the execution.
Specifications
  • Colour code:
    Light pink
  • Volume:
    10 - 100 µl
  • Description:
    P12×100M
  • Seq:
    09
  • Accuracy:
    ±0,25 - ±0,80
  • Imprecision:
    ≤0,14 - ≤0,25 µl
  • Recommended tips:
    D200 / DF100
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • TRACKMAN® Connected, EU
    TRACKMAN® Connected, EU
    613-5996
    Log in for availability
    1 * 1 Items - 2 031,00 €