Order Entry
Finland
ContactUsLinkComponent
Soil depth therm., similar DIN 58664, -10+30:0,1°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, built-in length 500mm, complete with plastic guide tube, protecting cap and packing ring, with thermom 1 * 1 items
Catalog # AMARK188450-CB
Supplier:  Amarell
Soil depth therm., similar DIN 58664, -10+30:0,1°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, built-in length 500mm, complete with plastic guide tube, protecting cap and packing ring, with thermom 1 * 1 items
Catalog # AMARK188450-CB
Supplier:  Amarell
Frequently Bought Together