Order Entry
Finland
ContactUsLinkComponent
Meteorological Extreme therm., minimum type, blue glass index, DIN 58653, -40+40:0,5C, with glass saddle and metal cap, milk glass scale, cap. lens front, blue reflecting, alcohol filled, 290mm long, diameter 18+/-0,5mm, government verified, with Ve 1 * 1 items
Catalog # AMARK173044-ALC/VC
Supplier:  Amarell
Meteorological Extreme therm., minimum type, blue glass index, DIN 58653, -40+40:0,5C, with glass saddle and metal cap, milk glass scale, cap. lens front, blue reflecting, alcohol filled, 290mm long, diameter 18+/-0,5mm, government verified, with Ve 1 * 1 items
Catalog # AMARK173044-ALC/VC
Supplier:  Amarell
Frequently Bought Together