Tissue Culture Flask, 75cm, Hydrophobic Surface (Susp), Canted Neck, Non-Vented Cap, Sterile,Non-Pyrogenic, Non-Cytotoxic, 5 Per Bag, Green 1 * 100 items
Catalog # SARS83.3911.500
Tissue Culture Flask, 75cm, Hydrophobic Surface (Susp), Canted Neck, Non-Vented Cap, Sterile,Non-Pyrogenic, Non-Cytotoxic, 5 Per Bag, Green 1 * 100 items
Catalog # SARS83.3911.500