Order Entry
Finland
Orders LinkContactUsLinkComponent
Hand wash, SURE®

Hand wash, SURE®

SURE® Hand Wash, 1,3 L
Catalog # 115-2955
   Safer Human and Environmental Health|Sustainable Materials
Supplier:   Diversey
Hand wash, SURE®
Catalog # 115-2955
Supplier:  Diversey
   Safer Human and Environmental Health|Sustainable Materials
Hand wash, SURE®
Hand wash, SURE®

SURE® Hand Wash, 1,3 L

Price
48,00 €
Log in for availability
1 * 4 Items
Product Details & Documents

Gentle and effective hand wash with a 100% natural perfume. Cleans hands leaving them hygienically fresh.

  • Free from parabens and other preservatives.
  • Contains plant-based actives from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product or user
  • 100% natural perfume with a refreshing lemon fragrance
  • Skin compatability tested (Patch test)

SURE® is a comprehensive offering of plant-based, cleaning products designed to deliver superior professional results while being safe for people and kind to the environment.

With environmentally-friendly ingredients from renewable resources, SURE® products meet the daily requirements for kitchens, facilities, and personal care.

Certifications: OECD 301B. Cradle to cradle certification at gold level.

Ordering information: 1.3 L cartridge is suitable for Diversey IntelliCare Dispenser (manual/hybrid).

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Documents

Specifications
  • Description:
    SURE® Hand Wash, 1,3 L
  • Seq:
    03
  • Cat. No.:
    115-2955
  • Environmental certification:
    OECD 301B
    Cradle to creadle certification at gold level
  • Environmentally Preferable:
    Safer Human and Environmental Health|Sustainable Materials