Order Entry
Finland
Orders LinkContactUsLinkComponent
4-Hydroxybenzaldehyde

4-Hydroxybenzaldehyde

Bottle, 1 kg
Catalog # APOSOR13187-1KG
Irritant
Supplier:   Apollo Scientific
4-Hydroxybenzaldehyde
Catalog # APOSOR13187-1KG
Supplier:  Apollo Scientific
CAS Number:  123-08-0
4-Hydroxybenzaldehyde

Bottle, 1 kg

CAS Number:  123-08-0
Price
47,10 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:123-08-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00006939
  • Molecular Weight:122.12 g/mol
  • Boiling Pt:191 °C/50mmHg
  • Melting Pt:112…116 °C
  • Density:1.129 g/cm³ (25 °C)
  • Flash Pt:174 °C
  • Storage Temperature:Ambient
  • EINECS:204-599-1

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.