Order Entry
Finland
Orders LinkContactUsLinkComponent
 

N-Methylmorpholine-N-oxide

Bottle, 25 g
Catalog # APOSOR323297-25G
Irritant
Supplier:   Apollo Scientific
N-Methylmorpholine-N-oxide
Catalog # APOSOR323297-25G
Supplier:  Apollo Scientific
CAS Number:  7529-22-8
N-Methylmorpholine-N-oxide

Bottle, 25 g

CAS Number:  7529-22-8
Price
22,10 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide
  • CAS Number:7529-22-8
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00005947
  • Molecular Weight:117.15 g/mol
  • Melting Pt:180…184 °C
  • Storage Temperature:Refrigerator
  • EINECS:231-391-8

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications