Order Entry
Finland
Orders LinkContactUsLinkComponent
Heller's basal salt mixture

Heller's basal salt mixture

Heller's basal salt mixture 10 L, 10 L
Catalog # APOSPMH393-10L
Supplier:   Apollo Scientific
Heller's basal salt mixture
Catalog # APOSPMH393-10L
Supplier:  Apollo Scientific
CAS Number:  
Heller's basal salt mixture

Heller's basal salt mixture 10 L, 10 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents

Contains macro/micronutrients as described by Heller, 1953 Plant tissue culture testedFull specification given in appendix.

  • MFCD00283413
Specifications

Specifications

  • Pk:
    10 L
  • Description:
    Heller's basal salt mixture 10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.