4'-Fluoroacetophenone 99%
Bottle, 100 g
Catalog # APOSPC3380-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- CAS Number:403-42-9
- Formula:C₈H₇FO
- MDL Number:MFCD00000354
- Molecular Weight:138.14 g/mol
- Boiling Pt:196 °C
- Density:1.138 g/cm³ (25 °C)
- Flash Pt:71 °C
- Storage Temperature:Ambient
- EINECS:206-960-9
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



