Order Entry
Finland
Orders LinkContactUsLinkComponent
3,4-Dichlorophenol 98%

3,4-Dichlorophenol 98%

Bottle, 250 g
Catalog # APOSOR0058-250G
Irritant
Corrosive
Environment
Supplier:   Apollo Scientific
3,4-Dichlorophenol 98%
Catalog # APOSOR0058-250G
Supplier:  Apollo Scientific
CAS Number:  95-77-2
3,4-Dichlorophenol 98%

Bottle, 250 g

CAS Number:  95-77-2
Price
228,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:95-77-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002258
  • Molecular Weight:163 g/mol
  • Boiling Pt:145…146 °C
  • Melting Pt:64…68 °C
  • Storage Temperature:Ambient
  • EINECS:202-450-5
  • UN:2020
  • ADR:6.1,III
  •  :Density: 1.6 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.