Order Entry
Finland
Orders LinkContactUsLinkComponent
3,4-Dichlorophenol 98%

3,4-Dichlorophenol 98%

Bottle, 100 g
Catalog # APOSOR0058-100G
Irritant
Corrosive
Environment
Supplier:   Apollo Scientific
3,4-Dichlorophenol 98%
Catalog # APOSOR0058-100G
Supplier:  Apollo Scientific
CAS Number:  95-77-2
3,4-Dichlorophenol 98%

Bottle, 100 g

CAS Number:  95-77-2
Price
145,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:95-77-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002258
  • Molecular Weight:163 g/mol
  • Boiling Pt:145…146 °C
  • Melting Pt:64…68 °C
  • Storage Temperature:Ambient
  • EINECS:202-450-5
  • UN:2020
  • ADR:6.1,III
  •  :Density: 1.6 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications