Order Entry
Finland
Orders LinkContactUsLinkComponent
Mouse recombinant Acrp30

Mouse recombinant Acrp30

1 mg
Catalog # APOSBITP1001-1MG
Supplier:   Apollo Scientific
Mouse recombinant Acrp30
Catalog # APOSBITP1001-1MG
Supplier:  Apollo Scientific
CAS Number:  
Mouse recombinant Acrp30

1 mg

CAS Number:  
Price
6 810,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1001-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Acrp30
  • Protein synonyms:
    Adiponectin
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.