Order Entry
Finland
Orders LinkContactUsLinkComponent
4'-Fluoroacetophenone 95%

4'-Fluoroacetophenone 95%

Bottle, 1 kg
Catalog # APOSPC2887-1KG
Irritant
Supplier:   Apollo Scientific
4'-Fluoroacetophenone 95%
Catalog # APOSPC2887-1KG
Supplier:  Apollo Scientific
CAS Number:  403-42-9
4'-Fluoroacetophenone 95%

Bottle, 1 kg

CAS Number:  403-42-9
Price
73,60 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
  • CAS Number:403-42-9
  • Formula:C₈H₇FO
  • MDL Number:MFCD00000354
  • Molecular Weight:138.14 g/mol
  • Boiling Pt:194…196 °C
  • Melting Pt:4 °C
  • Density:1.138 g/cm³ (25 °C)
  • Flash Pt:71 °C
  • Storage Temperature:Ambient
  • EINECS:206-960-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.