Order Entry
Finland
Orders LinkContactUsLinkComponent
 

L(-)-Mannose

Bottle, 1 g
Catalog # APOSBIFS0112-1G
Irritant
Supplier:   Apollo Scientific
L(-)-Mannose
Catalog # APOSBIFS0112-1G
Supplier:  Apollo Scientific
CAS Number:  10030-80-5
L(-)-Mannose

Bottle, 1 g

CAS Number:  10030-80-5
Price
206,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
  • CAS Number:10030-80-5
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00136021
  • Molecular Weight:180.16 g/mol
  • Melting Pt:129…131 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.