Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Fluorescence Polarization Filter with holder 590nm BW 20nm Polarized Vertical 1 * 1 items
Catalog # MLDV6590-0104
Supplier:
MOLECULAR DEVICES
Expand image
Fluorescence Polarization Filter with holder 590nm BW 20nm Polarized Vertical 1 * 1 items
Catalog # MLDV6590-0104
Supplier:
MOLECULAR DEVICES