0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Fluorescence Polarization Filter with holder 465nm BW 35nm Polarized Vertical 1 * 1 items
MLDV6590-0091
:
MOLECULAR DEVICES
Fluorescence Polarization Filter with holder 465nm BW 35nm Polarized Vertical 1 * 1 items
MLDV6590-0091
:
MOLECULAR DEVICES