Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Thermo Scientific TSX ULT Freezer, -86C, 400 Box Capacity, 230V/50 Hz 1 * 1 items
Catalog # REVCTSX40086V
Supplier:
REVCO TECHNOLOGIES
Expand image
Thermo Scientific TSX ULT Freezer, -86C, 400 Box Capacity, 230V/50 Hz 1 * 1 items
Catalog # REVCTSX40086V
Supplier:
REVCO TECHNOLOGIES