Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cogent HPS Cyano HPLC Column 120A 5um High Purity Silica. 21.2mm ID x 100mm. 1 * 1 items
Catalog # MICS75025-P100
Supplier:
MICROSOLV
Expand image
Cogent HPS Cyano HPLC Column 120A 5um High Purity Silica. 21.2mm ID x 100mm. 1 * 1 items
Catalog # MICS75025-P100
Supplier:
MICROSOLV