Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ICP Standard Chromium 1,000µg/ml in 2 - 5% HNO3 (v/v) 1 * 500 mL
Catalog # REGEPCR2C2
Supplier:
REAGECON
Expand image
ICP Standard Chromium 1,000µg/ml in 2 - 5% HNO3 (v/v) 1 * 500 mL
Catalog # REGEPCR2C2
Supplier:
REAGECON