Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Additional S/S Wire Basket - Stainless steel mesh basket for Top Loading QCS Priorclaves. 210mm Deep x 450mm dia. (2x 100L, 3 x 150L) 1 * 1 items
PRIOPC/HDB/000
:
Priorclave
Additional S/S Wire Basket - Stainless steel mesh basket for Top Loading QCS Priorclaves. 210mm Deep x 450mm dia. (2x 100L, 3 x 150L) 1 * 1 items
PRIOPC/HDB/000
:
Priorclave