Order Entry
Finland
ContactUsLinkComponent
Tubing threaded 40x300x3.2mm gl45 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.048
Supplier:  witeg Labortechnik
Tubing threaded 40x300x3.2mm gl45 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.048
Supplier:  witeg Labortechnik
Frequently Bought Together