Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TARGET SCREW THREAD SOLID TOP, POLYURETHANE WHITE CAP, PTFE/PE LINER, 100/PK 1 * 100 items
Catalog # NSCAC4010-99
Supplier:
THERMO NATIONAL SCIENTIFIC
Expand image
TARGET SCREW THREAD SOLID TOP, POLYURETHANE WHITE CAP, PTFE/PE LINER, 100/PK 1 * 100 items
Catalog # NSCAC4010-99
Supplier:
THERMO NATIONAL SCIENTIFIC