PAH-Mix 9, each µg/mL in Acetonitrile, 10 mL Dibenzo[a,h]anthracen 10,0 µg/ml Benzo[a]anthracen 20,0 µg/ml 6-Methylchrysen 20,0 µg/ml Benzo[e]pyren 20,0 µg/ml Benzo[j]fluoranthen 20,0 µg/ml Benzo[k]fluoranthen 20,0 µg/ml Benzo[a]pyren 20,0 µg/ml Benz 1 * 10 ml
Catalog # PROOSL24213
Supplier: LGC Standards PROMOCHEM