Order Entry
Finland
ContactUsLinkComponent
Air Flow:163 m 3/h Vacuum Pressure:2515 mmH2O Volt:120 V Ampere:8.3 A Watt:1000 W. Hertz:60 Hz. Dry Recovery: 15 L Dry Recovery: 15 L Collection Bag Capacity: 7.6 L Noise Level: 60 dB Dimensions: H56cmW41cmL41c
Catalog # TIGV110762AP
Supplier:  TIGER-VAC
Air Flow:163 m 3/h Vacuum Pressure:2515 mmH2O Volt:120 V Ampere:8.3 A Watt:1000 W. Hertz:60 Hz. Dry Recovery: 15 L Dry Recovery: 15 L Collection Bag Capacity: 7.6 L Noise Level: 60 dB Dimensions: H56cmW41cmL41c
Catalog # TIGV110762AP
Supplier:  TIGER-VAC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.