Order Entry
Finland
ContactUsLinkComponent
EACH COMPRESSOR COMES ASSEMBLED ON AN INTERNALLY AND EXTERNALLY COATED AIR TANK AND INCLUDES A SELF-PURGING REGENERATIVE DRYER WITH AFTERCOOLER. ONE VERSION INCLUDES A SOUND ENCLOSURE AND HAS A SOUND LEVEL OF LESS THAN 60 DB. MODELS IN 115 V - 60 HZ 1 * 1 items
Catalog # PERCN0777881
Supplier:  PerkinElmer
EACH COMPRESSOR COMES ASSEMBLED ON AN INTERNALLY AND EXTERNALLY COATED AIR TANK AND INCLUDES A SELF-PURGING REGENERATIVE DRYER WITH AFTERCOOLER. ONE VERSION INCLUDES A SOUND ENCLOSURE AND HAS A SOUND LEVEL OF LESS THAN 60 DB. MODELS IN 115 V - 60 HZ 1 * 1 items
Catalog # PERCN0777881
Supplier:  PerkinElmer
Frequently Bought Together