Order Entry
Finland
ContactUsLinkComponent
Goat anti-Mouse IgG (H&L), F(ab)'2 fragment, FITC conjugated, min. cross-reactivity to bovine, goat, human, rabbit, or rat IgG 1 * 0,5 mg
Catalog # AGRIAS163452
Supplier:  Agrisera
Goat anti-Mouse IgG (H&L), F(ab)'2 fragment, FITC conjugated, min. cross-reactivity to bovine, goat, human, rabbit, or rat IgG 1 * 0,5 mg
Catalog # AGRIAS163452
Supplier:  Agrisera