Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Microplate reader, Gemini™ EM

Gemini™ EM microplate reader, multi-mode, with current version of SoftMax® Pro and 1 additional year MRC service plan covering parts and labour
Catalog # MLDVEM-SC-ACAD
Supplier:   MOLECULAR DEVICES
Microplate reader, Gemini™ EM
Catalog # MLDVEM-SC-ACAD
Supplier:  MOLECULAR DEVICES
Microplate reader, Gemini™ EM
Microplate reader, Gemini™ EM

Gemini™ EM microplate reader, multi-mode, with current version of SoftMax® Pro and 1 additional year MRC service plan covering parts and labour

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Gemini™ EM microplate spectrofluorometer provides a flexible environment to determine the optimal excitation and emission settings for most fluorescent intensity assays. The Gemini EM reader with dual monochromators allows researchers to try new and novel dyes without having to purchase expensive filter sets.

  • Measure fluorescence from solution- or cell-based assays by switching between top and bottom fluorescence reading
  • Up to 4 wavelength pairs can be read for endpoint and kinetic measurements
  • Read a variety of sample formats from 6- to 384-well microplates in endpoint, kinetic, spectral scan, and well scan modes
  • Monitoring of temperature-sensitive reactions with consistent temperature regulation from ambient +4 to 45 °C
  • Thorough mixing of samples in microplate wells by user-programmable Automix feature
  • Automate seamlessly with the StakMax® microplate handling stacker (available separately)

The reader uses two scanning monochromators to determine the optimal excitation and emission settings for the spectral characteristics of fluorophores when used in an assay. If the best wavelengths for a fluorophore of interest are unknown, the Gemini EM reader can scan both excitation and emission spectra and determine the optimal settings.

 : Supplied with SoftMax® Pro data acquistion and analysis software.

Specifications
  • Description:
    Gemini™ EM microplate reader, multi-mode, with current version of SoftMax® Pro and 1 additional year MRC service plan covering parts and labour
  • Seq:
    02
  • Incubation range:
    Ambient +4 °C to 45 °C
  • Light source:
    Xenon flash lamp (1 joule/flash)
  • Temperature uniformity:
    <1 °C at 37 °C set point
  • Wavelength bandwidth:
    9 nm
  • Wavelength accuracy:
    ±2,0 nm
  • Weight:
    16 kg
  • Cat. No.:
    MLDVEM-SC-ACAD
  • Detector:
    Photomultiplier (R-3896)
  • Wavelength selection:
    1,0 nm increments
  • Model:
    Gemini™ EM
  • Wavelength range:
    250 - 850 nm (excitation), 360 - 850 nm (emission)
  • W×D×H:
    580×380×220 mm
  • Software:
    SoftMax® Pro
  • Measurement time:
    Reads a 96-well plate in <15 s, 384-well plate in <45 s
Complementary Products

Product Information

  • Catalog #
    Description
    Pk
    Serum format
    Sterility
    Packed
    Recommended working volume
    Culture area
    Sterile
    Colour
    Availability
    Price
  • 31028.
    AccuClear® Ultra high sensitivity dsDNA quantitation kit with 7 DNA standards
    1.000 Assays
    Log in for availability
    1 * 1 Each - 518,00 €

  • 31007.
    AccuBlue® Broad range dsDNA quantitation kit with 9 DNA standards
    1.500 Assays
    Log in for availability
    1 * 1 Each - 435,00 €

  • 30002.
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 KIT
    Log in for availability
    1 * 1 Each - 440,00 €

  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 Each - 417,00 €

  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    15
    Log in for availability
    1 * 500 mL - 322,00 €

  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 Each - 294,00 €

  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 Each - 631,00 €

  • 31006.
    AccuBlue® High sensitivity dsDNA quantitation kit with 8 DNA standards
    1.000 Assays
    Log in for availability
    1 * 1 Each - 435,00 €

  • 734-2327
    1/bag
    0,075 - 0,20 ml
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 Items - 185,00 €

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • SpectraDrop™ HTS kit
    SpectraDrop™ HTS kit
    MLDV0200-6267
    Log in for availability
    1 * 1 Items

Product Information

  • Catalog #
    Availability
    Price
  • SpectraDrop™ 24-well kit
    SpectraDrop™ 24-well kit
    MLDV0200-6262
    Log in for availability
    1 * 1 Items