Order Entry
Finland
ContactUsLinkComponent
BioSpectrum 615, MOTORISED fixed 25mm, f/0.95, MOTORISED Lift615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, with binning capability (3.2 Mp). Peltier cooling to -60C from ambient. Camera mount and 25mm, f/ 1 * 1 items
Catalog # UVPA97-0715-02
BioSpectrum 615, MOTORISED fixed 25mm, f/0.95, MOTORISED Lift615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, with binning capability (3.2 Mp). Peltier cooling to -60C from ambient. Camera mount and 25mm, f/ 1 * 1 items
Catalog # UVPA97-0715-02