MP ALUMINA G-TLC 1 * 1 kg
Catalog # ICNA0204409
Supplier: MP Biomedicals
Frequently Bought Together
Metanoli ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Vesi, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Serologinen pipetti, Standard Line
Supplier: VWR Collection
Käsineet, nitriili, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Asetonitriili ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Kasvopyyhkeet
Supplier: VWR Collection
VWR®, Käsineet, nitriili
Supplier: VWR Collection
pH-testiliuskat, MQuant®, Supelco®
Supplier: Merck
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Asetoni ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Käsineet, nitriili
Supplier: VWR Collection