Order Entry
Finland
Orders LinkContactUsLinkComponent
Polysorbate 40 NF

Polysorbate 40 NF

Glass, amber, 4 L
Catalog # SPCMPO134-4LTGL
Supplier:   Spectrum Chemical
Polysorbate 40 NF
Catalog # SPCMPO134-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  9005-66-7
Polysorbate 40 NF

Glass, amber, 4 L

CAS Number:  9005-66-7
Price
667,00 €
Log in for availability
1 * 4 L
Product Details & Documents
Synonyms:  Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
  • CAS Number:9005-66-7
  • Formula:C₆₂H₁₂₂O₂₆
  • MDL Number:MFCD00165345
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    4 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.