Polysorbaatti 40 NF
Poly drum, 20 L
Catalog # SPCMPO134-20LT
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Storage Temperature:Huoneenlämpö
Specifications
Specifications
- Pakkaustyyppi:Poly drum
- Pak.:20 L



