Order Entry
Finland
Orders LinkContactUsLinkComponent
Fumaric acid NF

Fumaric acid NF

Poly pail, 12 kg
Catalog # SPCMF1096-12KG
Irritant
Supplier:   Spectrum Chemical
Fumaric acid NF
Catalog # SPCMF1096-12KG
Supplier:  Spectrum Chemical
CAS Number:  110-17-8
Fumaric acid NF

Poly pail, 12 kg

CAS Number:  110-17-8
Price
5 420,00 €
Log in for availability
1 * 12 kg
Product Details & Documents

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:13,04308
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:287 °C
  • Density:1.625 g/cm³ (20 °C)
  • Flash Pt:273 °C
  • EINECS:203-743-0
Specifications

Specification Test Results

  • Assay:
    99.5% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg

Specifications