Order Entry
Finland
Orders LinkContactUsLinkComponent
Ethanolamine NF

Ethanolamine NF

Poly drum, 20 L
Catalog # SPCMM1322-20LT
Corrosive
Irritant
Supplier:   Spectrum Chemical
Ethanolamine NF
Catalog # SPCMM1322-20LT
Supplier:  Spectrum Chemical
CAS Number:  141-43-5
Ethanolamine NF
Ethanolamine NF

Poly drum, 20 L

CAS Number:  141-43-5
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:13,03762
  • Boiling Pt:171 °C (1013 hPa)
  • Melting Pt:10.5 °C
  • Density:1.0164 g/cm³ (20 °C)
  • Flash Pt:93 °C
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pack type:
    Poly drum
  • Pk:
    20 L

Specifications