Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Bottom sieve SK 300, stainless steel, trapezoid holes 0.50 mm 1 * 1 items
Catalog # RETS02.407.0101
Supplier:
Retsch GmbH
Expand image
Bottom sieve SK 300, stainless steel, trapezoid holes 0.50 mm 1 * 1 items
Catalog # RETS02.407.0101
Supplier:
Retsch GmbH