Order Entry
Finland
ContactUsLinkComponent
Phosphoenolpyruvate carboxylase (PEPC EC 4.1.1.31) serves as an important control element in the regulation of photosynthetic carbo n metabolism in C4 and CAM plants. 1 * 100 µl
Catalog # AGRIAS09458S
Supplier:  Agrisera
undefined
Phosphoenolpyruvate carboxylase (PEPC EC 4.1.1.31) serves as an important control element in the regulation of photosynthetic carbo n metabolism in C4 and CAM plants. 1 * 100 µl
Catalog # AGRIAS09458S
Supplier:  Agrisera