Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
2,000UG/ML FE 5000UG/ML AL, CA, MG IN 20% HCL, 250ML 1 * 250 mL
PROOVHG-ICSA-250
:
LGC Standards PROMOCHEM
2,000UG/ML FE 5000UG/ML AL, CA, MG IN 20% HCL, 250ML 1 * 250 mL
PROOVHG-ICSA-250
:
LGC Standards PROMOCHEM